PG2 DOT-Compliant Sharps Disposal Containers, Covidien
Supplier: Covidien
8998PG2 8997PG2 8998PG2 8997PG2 8998SPG2 8930SA 8998SPG2 8936PG2
89002-264CS 698.92 CAD
89002-264 89002-266 76480-332 93000-308 CA89002-264 89004-532 CA89002-266 CA89004-532
PG2 DOT-Compliant Sharps Disposal Containers, Covidien
Bins Sharps Containers
These PG2-rated sharps disposal containers are ideal for the transport of sharps, cultures, and stocks
Container labeling is DOT-compliant, eliminating the need for secondary packaging when transporting by a private or contract carrier. Containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.
An absorbent pad helps contain excess fluid. The lid gasket provides leak resistance. Molded carrying handles allow easy transport