BETA-GARD™ Waste Containers, IBI Scientific
Supplier: IBI Scientific
WH-75 WH-160 WH-250 FL-60 FL-150 WH-280-M WH-280-M
13500-534EA 436.78 CAD
13500-534 13500-536 13500-538 13500-540 13500-542 IBWH-280-M 490012-996
BETA-GARD™ Waste Containers, IBI Scientific
Bins Waste Bins
Containers for the disposal of radioactive waste can also be used with disposable NRC-approved waste bags
Constructed of durable 13mm (1/2") cast acrylic for maximum protection. Available with hinged or flip-top lids. Container IBWH-280-M has a handle. Containers with flip-top lids have a 2cm hole for disposal of contaminated pipets.