Nalgene® Diagnostic Bottles, PETG, Sterile, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
342035-0005 342035-0020 342035-0010 2035-0010 2035-0020 2035-0005
73520-082CS 357.71 CAD
73520-082 73520-084 73520-080 73520-086 73520-078 73520-088
Nalgene® Diagnostic Bottles, PETG, Sterile, Thermo Scientific
Bottles Plastic Bottles
PETG single-use diagnostic bottles are ideal for sampling, storage, and shipment of reagents and buffer solutions.
  • Bottles and caps are gamma sterilized to 10⁻⁶ sterility assurance levels and have a lot number
  • Nonpyrogenic, noncytotoxic
Ordering information: Supplied with white, lined screw caps with a 20-415 thread finish. Tray-packed bottles are packed 20 bottles per tray, 5 trays per case. Bulk-packed bottles are packed 100 bottles per tray, 5 trays per case. All trays are packaged in double-lined cartons for cleanroom use, with caps assembled.