Oily Waste Cans, Justrite®
Supplier: Justrite
09500 09100 09700 09300 9310 9510 9710
56609-762EA 377 CAD
56609-762 56610-539 56610-540 56610-541 10836-818 10836-820 10836-822
Oily Waste Cans, Justrite®
Bins Waste Bins
Essential wherever solvent cloths and wiping rags are used
  • Can be operated by hand or foot-lever opening device
  • Lid closes automatically when not in use
  • Round construction and elevated bottom encourage circulation of air
  • One-piece cover facilitates use of collection bags
  • Steel construction with powder coat paint finish
When operated by hand or foot lever opening device, the cover opens to a 60° angle and closes automatically when released. Round construction and elevated bottom encourage circulation of air around can to disperse heat and preventMore Product Information