Oily Waste Cans, Polyethylene, Eagle Manufacturing
Supplier: Eagle Manufacturing
935FL 933FLY 935FLY 933FL 937FL 937FLY
56610-175EA 459.76 CAD
56610-175 56610-216 56610-218 56617-330 56617-335 56610-220
Oily Waste Cans, Polyethylene, Eagle Manufacturing
Bins Waste Bins
Cans are designed for the disposal of solvent and oily waste, rags, and other materials subject to spontaneous combustion.
  • Impact-resistant
  • Corrosion-resistant
  • Rustproof
Impervious to many liquid chemicals, solvents, and corrosives. A foot lever raises the lid which closes automatically after use.
FM approved.