SatPax® 1000, Presaturated Wipers, Berkshire
Supplier: Berkshire
SPX100000212 SPXC10000212 SPXC1000112R SPX100000112 SPX1000.011.12
21912-104CS 415.41 CAD
21912-104 69999-302 69999-306 21912-102 76498-148
SatPax® 1000, Presaturated Wipers, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SatPax® wipers provide a convenient combination of solvent and wiper that improves the cleaning process.It resolves safety issues associated with storage, mixing, and dispensing of solvents.
  • Low extractables and fiber and particle counts
  • Smooth and durable with good wet strength
  • Reduces alcohol usage and preparation/handling costs
  • Increases cleaning efficiency
SatPax® 1000 nonwoven polyester/cellulose wipers are saturated with semiconductor grade isopropyl alcohol and deionized water. Knitted polyester wipers are cleanroom laundered and areMore Product Information