TechniSat® Presaturated Wipers, Texwipe®
Supplier: Texwipe
TX8488 TX1041 TX1045 TX1065 TX1067 TX1065
TWTX8488CS 559.64 CAD
TWTX8488 CATW-TX1041 CATWTX1045 CATWTX1065 TWTX1067 TW-TX1065
TechniSat® Presaturated Wipers, Texwipe®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These convenient wipers provide portion control and reduced potential of spillage in and out of the cleanroom.
  • Fibers are blended by a hydroentanglement procedure
  • Uses water instead of chemicals to bind
  • Excellent bidirectional strength and a very low extractable level
  • Wipers are controlled for traceability
The blended material of 55% cellulose and 45% polyester provides the high absorbency of a natural fiber with the cleanliness and strength of a synthetic material. For non sterile wipers, the 70% semiconductor-grade isopropyl alcohol is filtered to 0.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable