Hybridization Bottles, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
805000 805001 805003 805007 805011 805020 805027
16300-300EA 108.85 CAD
16300-300 16300-302 16300-304 16300-306 16300-308 16300-312 16300-314 16300-316 16300-318 16300-320
Hybridization Bottles, WHEATON®, DWK Life Sciences
Bottles Hybridisation Bottles/Tubes
These bottles are ideal for nucleic acid and protein blotting procedures that use a minimum of probe solution.
These borosilicate bottles are produced with a heavy wall for added safety and are available with a slip-resistant safety coating. The 35 mm bottles incorporate a fluoropolymer resin/silicone lined cap to prevent leakage. Both bottles incorporate a polypropylene cap with a welded fluoropolymer resin/silicone liner. Optional acrylic storage rack holds up to six hybridization bottles.
They are a safe and efficient alternative to messy bags, and allow hybridization and wash solutions to be quickly poured in and out. Bottles fit most popular hybridization ovens to simplify procedures such as Southern, Northern, Western, and slot blots.