Synthware Gas Washing Bottle with Fritted Disc and Two Hose Connections, Kemtech America
Supplier: Kemtech America
B300125C B300125X B300250C B300500C B300500X
60000-728EA 470.64 CAD
60000-728 60000-730 60000-732 60000-736 60000-738
Synthware Gas Washing Bottle with Fritted Disc and Two Hose Connections, Kemtech America
Bottles Gas Washing Bottles
Gas washing bottle with [ST]45/50 cap-style stopper and fritted disc angled at 15° for even distribution of gas throughout absorbing material.
  • Heavy wall borosilicate construction
  • Fritted disc
  • Large base
  • 45/50 ground glass joint
Available with a coarse or extra-coarse fritted disc. Springs (60000-758) are ordered separately.