ESD Safe Dispenser Flux Bottle with Needle, R & R Lotion
Supplier: R & R Lotion
FD-1-ESD FD-2-ESD
56614-132EA 34.84 CAD
56614-132 56614-134 56614-152
ESD Safe Dispenser Flux Bottle with Needle, R & R Lotion
Bottles Dropping Bottles
ESD safe flux bottles are available in 20, and 26 guage needle sizes.
  • Designed for dispensing solvents and fluxes
  • Polyethylene construction
R&R Lotion I.C. antistatic dissipative ESD safe flux bottles are available in 20, and 26 guage needle sizes. Cleanroom blue static dissipative containers have an average surface resistivity of 10⁹ to 10¹⁰. All blue solvent dispensers will dissipate a charge of +/- 5000 volts in less than 2 seconds at 40 percent relative humidity. By their chemical nature, they will not have any chemical reactivity withMore Product Information