Durable Insulated Containers, Less than 5 cu. ft., Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
390 301 450 303 422 300
16191-311EA 1422.46 CAD
16191-311 35742-100 35742-101 35742-104 35742-417 73320-006 89496-028 89496-030
Durable Insulated Containers, Less than 5 cu. ft., Sonoco ThermoSafe
Coolers Dry Ice Chests
Thermosafe Storage chests offer an excellent solution for the storage and transportation of temperature sensitive specimens for labs and universities or for the collection of field samples.
  • No power source needed so can be placed where you need it for added safety and convenience
  • Smooth polyethylene interior and exterior makes it easy to wipe clean
  • Rounded internal corners ensure the inside can be thoroughly cleaned too
  • Polyurethane insulation between the interior and exterior walls provides exceptional insulation to preserve the quality of samples or ice
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable