Bluesorb® 750 Wipers, Berkshire
Supplier: Berkshire
BS7500404.40 BS7500909.20 BS7501212.20
21914-707CS 1613.31 CAD
21914-707 21914-729 21914-740
Bluesorb® 750 Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
All-purpose Class 100 wipers are extremely absorbent.
  • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Blue color allows differentiation for guarding against migration into critical areas
  • Smooth, absorbent, and durable
  • Autoclavable
  • Not recommended for applications where 100% water is used; material may not be colorfast
Made of non-woven polyester/cellulose. Excellent dry and wet strength. Blue color provides an instant absorption indicator.