Cease-Fire® Waste Receptacles, Justrite®
Supplier: Justrite
26630G 26612G 26430
56610-547EA 802.57 CAD
56610-547 56610-553 56610-545
Cease-Fire® Waste Receptacles, Justrite®
Bins Waste Bins
For the safe accumulation of waste paper and other solvent-free combustibles. Contoured head directs smoke and gas from any fire inside the container back into the combustion area, cutting off the air supply and extinguishing flames in seconds. Provides round-the-clock protection against trash fires.
  • Specially contoured to direct smoke and gas from any fire inside the container, back into the combustion area, cutting off the air supply and extinguishing flames
  • Corrosion-free aluminum heads provide easy cleanup
  • Bodies are heavy-gauge steel with rust-preventative interior coating
For the safe accumulation of waste paper and other solvent-free combustibles
Aluminum heads are virtually corrosion-free, providing easy cleanup and long-lasting good looks. Steel heads are finished in a durable powder-coat paint finish and are recommended for applications where potentially abusive conditions make aluminum heads impractical. Drum bodies are heavy gauge steel, finished with a gray enamel exterior and a rust-preventive interior coating.
Available as a complete unit (drum and head), or heads and drums may be ordered separately. Head opening is 13.3cm (51/4") for 45.4 and 56.8L (12 and 15 gal.) receptacles, 17.8cm (7") for 113.6L (30 gal.) receptacles, and 20cm (77/8") for 208.2L (55 gal.) receptacles.
Contoured head directs smoke and gas from any fire inside the container back into the combustion area, cutting off the air supply and extinguishing flames in seconds. Provides round-the-clock protection against trash fires.
Certifications: Complete units are FM approved and UL listed. Separate heads are FM approved.