VITLAB® Dropping Bottles, LDPE, BrandTech
Supplier: Brandtech
94587 94687 94987 95087 95187
83009-066PK 245.11 CAD
83009-066 83009-068 83009-070 83009-072 83009-074
VITLAB® Dropping Bottles, LDPE, BrandTech
Bottles Dropping Bottles
Translucent dropping bottles come with a high-density polyethylene dropper screw cap.
  • Cut-to-size dropper with attached cap
  • LDPE
  • Five different sizes
The bottles provide excellent chemical resistance to aliphatic alcohols, aldehydes, alkalis, and acids.