Amplitude™ Epsilon™ Wipes, Contec®
Supplier: Contec
AMEP0001 AMEP0002
89030-174CS 637.75 CAD
89030-174 89030-176
Amplitude™ Epsilon™ Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Amplitude™ Epsilon™ Wipes are made of a special grade blue cellulose and polyester hydroentangled to form a wipe with improved cleanliness and excellent sorbency.
  • Recommended for ISO Class 5 to 6 environments
  • Dye-fast blue color allows for differentiation between wipes used in different areas
  • Double packaged to preserve product integrity
The cellulose uses a dye fast color, to minimize color bleeding. Blue color allows for differentiation in critical areas. Ideal for use in wet areas requiring spill pickup and cleanup.