Amplitude™ Delta™ Wipes, Contec®
Supplier: Contec
AMDE0001 AMDE0002 AMDE0003 AMDE0004
89030-156CS 1061.3 CAD
89030-156 89030-158 89030-160 89030-162
Amplitude™ Delta™ Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Amplitude™ Delta™ Wipes are made of 100% nonwoven polyester and manufactured using the spun lacing process with no binders or additives.
  • 100% nonwoven polyester is ideal for ISO Class 5 to 6 environments
  • Open fiber structure provides excellent sorptive capacity
  • Durable nonwoven polyester is low in both particles and extractables
  • 100% polyester is compatible with a wide range of solvents and disinfectants, including hypochlorites
  • Double packaged to preserve product integrity
A cost effective polyester wipe with excellent cleanliness levels. The open structure makes it highly sorbent. It is a very soft wipe with both good wet and dry strength. 100% polyester construction offers good chemical resistance and compatibility with most solvents and disinfectants.