Biohazard Waste Containers, Justrite®
Supplier: Justrite
05910 05930
56617-830EA 466.97 CAD
56617-830 56617-832
Biohazard Waste Containers, Justrite®
Bins Waste Bins
For temporary collection of hazardous biological waste
  • For segregation of infectious waste
  • Leakproof, sturdy metal construction
  • Simple, foot-operated, self-closing lid to reduce contamination
  • Biohazard bag is held securely around the circumference of the can
Caution: Not to be used with sharps.
May be used for temporary storage of contaminated laundry as well as other regulated waste.
Order Now
SPECIFICATIONS
Environmentally Preferable