Tweezers, Miniature, Excelta Corp®
Supplier: Excelta
M-00D M-2A-SA M-3-S M-4A-S M-4-S M-5-S M-5-SA M-AC M-H-S M-H-SA M-2-SA M-5-SA-ET M-00D-SA M-179A-RT M-3-CO-ET M-5-CO-ET M-5-S-ET M-159-RT M-159A-RT M-159B-RT M-159C-RT M-159D-RT M-159E-RT M-159G-RT M-159B-RTW M-159C-RTW M-159D-RTW M-759-RT M-759D-RT M-759-RTW M-759D-RTW
63043-440EA 195.07 CAD
63043-440 63043-448 63043-452 63043-454 63043-458 63043-464 63043-466 63043-472 63043-476 63043-478 94024-410 89411-848 89411-874 89411-910 76471-394 76471-396 76471-398 76471-400 76471-402 76471-404 76471-406 76471-408 76471-410 76471-412 76471-414 76471-416 76471-418 76471-420 76471-422 76471-424 76471-426 76471-428
Tweezers, Miniature, Excelta Corp®
Forceps
This 'short' tweezer is ideal for working under a scope.
  • Great for microscopy
  • Stainless steel construction-low oxidation
  • Variety of tip size/shape options
Miniature straight medium pointed tweezers are the most popular general purpose tweezers used with magnification. Their 'short' size makes them ideal for working under a scope. The 'ET' version features the Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat.
Order Now
SPECIFICATIONS
Environmentally Preferable