Nalgene® Heavy-Duty Polypropylene Carboys, Thermo Scientific
Supplier: Thermo Fisher Scientific
2226-0020 2226-0050
36494-090CS 1865.08 CAD
36494-090 36494-092
Nalgene® Heavy-Duty Polypropylene Carboys, Thermo Scientific
Bottles Carboys
Sturdy, thick-walled bottles are designed for heavy-duty applications.
  • Autoclavable
Use with optional quick filling/venting closures to create a vacuum trap. Quick filling/venting closures feature valved quick disconnect fittings, so that the carboy remains sealed when the tubulation fitting is removed.
Note: Before autoclaving, set cap on top of carboy without engaging threads.
Rugged caps with TPE gaskets guarantee leakproof service. Carboys can withstand repeated application of full vacuum for eight hour periods at 20 °C (68 °F).