Nalgene® Sanitary Carboys with Handles, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2630-0010 2630-0020 2630-0050
21020-262CS 667.78 CAD
21020-262 21020-264 21020-266
Nalgene® Sanitary Carboys with Handles, Polypropylene, Thermo Scientific
Bottles Carboys
Ideal for bioprocessing applications.
  • Convenient shoulder handles
Instead of a threaded closure system, carboys feature a 76 mm (3") sanitary flange that accepts standard TC fittings. Containers may be sealed securely with a solid end cap (16210-098) and clamp, or adapted to another TC fitting for filling and dispensing. A plastic true union clamp (16210-116) is recommended instead of a metal clamp.
Certifications: Meet USP-VI criteria and noncytotoxic.