UST Gasoline Surrogate Standards, Restek
Supplier: RESTEK CORP
30067
30082
30066
30068
RK30067EA
52.92
CAD
RK30067
RK30082
RK30066
RK30068
UST Gasoline Surrogate Standards, Restek
Standards
Petroleum Industry Standards
Standards are packaged in purge-and-trap-grade methanol in 1 mL ampoules.
- 2500 μg/mL in P&T methanol, 1 mL/ampoule
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).