KONTES® Fritted Hirsch Funnels, KIMBLE®, DWK Life Sciences
Supplier: DWK Life Sciences
KONTES®
955250-5023 955250-5044 955250-7544 955250-5026 955250-3523 955250-7523 955250-7526
KT955250-5044EA 207.83 CAD
KT955250-5044 KT955250-5023 KT955250-7523 KT955250-7544 KT955250-5026 KT955250-3523 KT955250-7526
KONTES® Fritted Hirsch Funnels, KIMBLE®, DWK Life Sciences
Funnels Filtration Funnels Filter Funnels
Funnels feature a conical shape that directs filtrate through a reduced-area frit to concentrate crystal residue for rewashing.