Diversified Biotech CoolSafe™ Systems
Supplier: Diversified Biotech
CSAF-1000 CSAF-2000 CSAF-3000 CSAF-3050 CSAF-3840 CSAF-4000 CSAF-5000 CSAF-9600
89024-860EA 2123.6 CAD
89024-860 89024-862 89024-864 89024-866 89024-868 89024-870 89024-872 89024-874
Diversified Biotech CoolSafe™ Systems
Coolers Benchtop Coolers
Anodized aluminum storage kits are ideal for sustaining cool or warm constant temperatures up to 10 hours during benchtop storage
Each kit includes one polystyrene CoolSafe storage box, one anodized aluminum cooling chamber rack, and two interchangeable cool bricks. Additional aluminum cooling chamber racks are also available.
Order Now
SPECIFICATIONS
Environmentally Preferable