Olympic™ Stacking Bins, Metro™
Supplier: Metropolitan Wire
SB90743CAS SB91055CAS SB91516CAS SB91055BAS SB90743BAS
CA22221-320EA 26.99 CAD
CA22221-320 CA22221-322 CA22221-324 CA22221-326 CA82002-514 CA82002-512 89207-068 89207-070
Olympic™ Stacking Bins, Metro™
Boxes ESD-Safe Boxes
Bins are durable, stackable, and feature high impact strength
  • A flat surface on the front of the bin is provided for easy placement of labels and bar codes
  • All bins have an open hopper for easy access to contents
  • Bins feature a lip on the edge of the rear panel for hanging
Choose from Benstat™ or Bentron™ bins:
Benstat is a static-dissipative polypropylene-based material with a surface resistivity of >105ohms/sq. and <1012ohms/sq. All Benstat bins are blue.
Bentron is a polypropylene-based material with a carbon additive to provide an electrically conductive path through the material. Bentron has a surface resistivity of <105ohms/sq. All Bentron bins are black.