VWR® Critical Print™ Cleanroom Combination Notebooks
Supplier: VWR International
P1114-10CG P1114-11CG
51127-500CS 558.13 CAD
51127-500 51127-502
VWR® Critical Print™ Cleanroom Combination Notebooks
Notebooks
Side-spiral notebooks are college ruled on the front of each page and have a 6.4 mm (1/4") grid on the back of each page.
  • Side-spiral design
  • Semi-transparent polyethylene cover
  • Bagged/wrapped twice
Low particle 28lb. bond paper. Semi-transparent, high-density polyethylene covers. Each notebook contains 50 sheets. Notebooks are Class 10 and are bagged/wrapped twice.