French Square Bottles, Wide Mouth
Supplier: Qorpak
GLC-01396 GLC-01345 GLC-01392 GLA-00832 GLA-00834 GLC-01363 GLA-00821 GLC-01307 GLC-01258 GLC-01283 GLC-01334 GLC-01387 GLC-01252 GLC-01272 GLC-01326 GLC-01354 GLC-01378 GLC-01377 GLA-00831 GLC-01341 GLC-01268 GLC-01320 GLC-01347 GLC-01241 GLC-01260 GLC-01311 GLC-01336 GLC-01364 GLA-00835 GLC-01255 GLC-01332 GLC-01331 GLC-01360 GLA-00829 GLC-01373 GLA-00823 GLC-01325 GLC-01370 GLC-01266 GLC-01318 242348 242349 242291 242346 242347 GLA-00826 GLC-01293 GLC-01284 GLC-01297 GLC-01298 GLC-01281 GLA-00830 GLA-00833
11311-240CS 448.02 CAD
11311-240 11311-344 11311-376 11311-504 11311-508 11311-656 11311-658 16085-227 16087-002 16151-408 16151-410 16151-996 16151-998 16152-010 16152-016 16152-188 16152-192 16188-027 16188-038 16188-049 16188-050 16188-082 16188-107 16188-118 16188-129 16188-130 16188-344 16188-346 16188-504 16188-526 16188-548 16188-560 16188-581 16188-708 16188-741 16188-854 16188-899 16189-008 16189-030 16189-096 16189-121 16189-154 16195-850 36319-187 36319-270 36319-271 36319-274 36319-279 36319-281 36319-282 36319-285 36319-288 89176-984 93001-634 16188-876 16807-004 89201-534 89204-870 89204-876 89204-878 30618-014 76170-356 76170-358 76170-416 76170-418 76170-420 76209-728 76209-730 76209-732 76209-734 76209-736 76209-738 89347-981 76419-772 76419-774 76419-776 76419-778 76419-780 76419-782 76419-784 76419-786 76419-788 76419-790 76419-792 76520-566 76539-348
French Square Bottles, Wide Mouth
Bottles Glass Bottles
Clear, flint glass French square bottles provide maximum content visibility.
  • Space saving design saves on shelf and storage space
The wide mouth opening is ideal for mixing, storing, and sampling.
Ordering information: Bottles 16152-188 and 16152-192 are supplied Ultra Cleaned. Bottles 16151-410, 36319-270, 36319-279, 36319-281, 36319-285, and 36319-288 are supplied pre-cleaned for use with volatiles.