VWR® CryoCoolers
Supplier: VWR International
414004-285 414004-286 414004-287
414004-285CS 3448.59 CAD
414004-285 414004-286 414004-287
VWR® CryoCoolers
Coolers Benchtop Coolers
Coolers hold 0.5 or 1.5 mL microcentrifuge tubes at or below -20°C (-4°F) for up to one hour on the benchtop or in transport.
  • Printed grid
  • Keyed lid
  • Nonskid feet
Cooler and lid are filled with nontoxic gel. Store at -25°C (-13°F).
Parts # 414004-287 and 414004-285 come with a locking handle.