Sample Bottles, Wide Mouth, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
225532 225534 225535 225536 225543 225546
66015-010CS 476.75 CAD
66015-010 66015-020 66015-042 66015-064 66015-166 66015-168 66015-224
Sample Bottles, Wide Mouth, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Sample bottles are designed for collecting, storing and exhibiting a wide variety of specimens in the lab or field.
  • Manufactured from Wheaton 800 soda-lime flint glass
PE caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suited for hygroscopic materials.
Packaging: Packed in convenient Lab File® cases.