AC Round Bottles, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W216995 W216996 W216997 W216998 W216999 W217000 W217001 W217002 W217004 W217005 W217006 W217007 W217008 W217009 W217003
14236-002CS 271.63 CAD
14236-002 14236-004 14236-006 14236-008 14236-010 14236-012 14236-014 14236-016 14236-020 14236-022 14236-024 14236-026 14236-028 14236-030 14236-018
AC Round Bottles, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Atlantic City round bottles are ideal for sampling use and can also be used to store dry powders, soil samples, and specimens.
  • Manufactured from Wheaton '800' glass
Certifications: Meets USP Type III standards.