PCR-Cooler and PCR Racks
Supplier: Eppendorf
022510541 022510525 022510509 0030124545
CA62111-758EA 178.78 CAD
CA62111-758 CA62111-760 CA62111-762 CA89135-368
PCR-Cooler and PCR Racks
Coolers Benchtop Coolers
Eppendorf PCR-Racks for safe PCR sample storage/transport with color coding for easy identification - ideal for 0.1, 0.2, 0.5 ml tubes/strips. The PCR-Cooler keeps samples cool for 1hr+ post 2hrs in -20 °C, with temperature indicator for sample safety.
  • Handling system for sample set-up, protection, transport, and storage of sensitive samples - keeps your samples safe
  • Clear temperature indicator: Color of PCR-Cooler changes when temperature exceeds 7 °C
  • Accommodates PCR vessels as tubes, strips, or plates for flexible vessel usage
  • Dry incubation technology reduces contamination risk of samples
  • Keeps an entire 96-well PCR plate cold for more than an hour at 0 °C (with two-hour precooling at –20 °C) for safe samples
Coolers for cool samples throughout PCR setup and racks for safe sample transport and storage.