MicroMop™ Flat String Mop Heads, Micronova
Supplier: Micronova
QDMH-1 MHC-1 MH-1 CRM-2 CRM-66
68400-202EA 151.96 CAD
68400-202 94023-782 68400-001 68400-002 68400-210 68400-778
MicroMop™ Flat String Mop Heads, Micronova
Mops
A selection of highly effective, flat-strand mops with high absorbency and low particulates. Available in three materials for specialized cleaning; polyvinyl alcohol, synthetic chamois, and die-cut polyester.
  • High absorbency and low particulates
  • Dry, residue-free cleaning results
  • Designed for cleanroom and controlled environments
Fabric options include:
PVA - non-woven polyvinyl alcohol with 600% absorbency and 10-12 second dry time; ideal for semiconductor and microelectronic cleaning applications
NovaLite™ - absorbent synthetic chamois; excellent fineMore Product Information