Weitlaner Retractors, Blunt, Sklar®
Supplier: Sklar
42-1875 22-8355 22-8345 42-1895 42-1865
82030-218EA 869.29 CAD
82030-218 82030-148 82030-222 82030-146 82030-220
Weitlaner Retractors, Blunt, Sklar®
Retractors
Sklar's® Weitlaner Retractor is a self-retaining, finger ring retractor with a cam ratchet lock used for holding back tissue and exposing a surgical site. It has curved shanks that lead to blades that have between two and six, sharp or blunt, outward curving prongs. The prongs interdigitate when the instrument is closed.
The Weitlaner retractor is a popular instrument, most commonly used in basic plastic surgery, large bone and joint procedures, and mastoid surgeries.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.