Safety-Coated Packer Bottles, Wide Mouth, Amber, Qorpak
Supplier: Qorpak
GLC-02294 GLC-02295 GLA-00964 GLA-00968 GLC-02285 GLC-02289 GLC-02283 GLC-02286 GLC-02290 GLC-07022 GLC-07023 GLC-07024 GLA-07053 GLC-14181 GLC-14183
11311-176CS 1055.26 CAD
11311-176 16085-028 16180-880 16180-883 16180-886 16180-892 16180-895 16180-903 16180-904 16180-906 89096-730 89096-732 89096-734 89096-736 89096-738 89096-740 93000-914 93000-916 93000-918 93000-920 93001-740 89212-384 89212-386 76170-394
Safety-Coated Packer Bottles, Wide Mouth, Amber, Qorpak
Bottles Glass Bottles
Reduce breakage and lab mishaps with these safety coated amber wide mouth packer bottles.
  • Safety-coated packer bottles are made of glass
If the glass breaks, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal.
Ordering information: Bottles 89096-730, 89096-732, 89096-734, 89096-736, 89096-738, and 89096-740 are supplied Ultra Cleaned. Bottles 93000-914, 93000-916, 93000-918, 93000-920, 89212-384, and 89212-386 are supplied vacuumed and ionized.