Weitlaner Retractors, Sharp, Sklar®
Supplier: Sklar
22-8245 42-1965 42-1975 42-1995
82030-142EA 809.69 CAD
82030-142 82030-224 82030-226 82030-228
Weitlaner Retractors, Sharp, Sklar®
Retractors
Sklar's® Weitlaner Retractor is a self-retaining, finger ring retractor with a cam ratchet lock used for holding back tissue and exposing a surgical site. It has curved shanks that lead to blades that have between two and six, sharp or blunt, outward curving prongs. The prongs interdigitate when the instrument is closed.
The Weitlaner retractor is a popular instrument, most commonly used in basic plastic surgery, large bone and joint procedures, and mastoid surgeries
Censitrac Ready products are industry standard and constructed of stainless steel.
It is also useful with small, deepMore Product Information