AC Medium Rounds, Clear, Wide Mouth, Graduated, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
5810133V-21 5810238V-21 5810448V-21 5810133C-21 5810238C-21 5810448C-21 5810858C-21 5811670C-21 5810448C-22 5810858C-22 5811670C-22 5810448C-23 5810858C-23 5811670C-23 5810133C-24 5810238C-24 5810133C-25 5810238C-25 5810133V-26 5810238V-26 5810448V-26 5810858V-26 5811670V-26 5810133C-26 5810238C-26 5810448C-26 5810858C-26 5811670C-26 5810238B 5810448B 5810858B 5811670B 5810858V-21 5811670V-21
89046-472CS 247.8 CAD
89046-472 89046-474 89046-476 89046-482 89046-484 89046-486 89046-488 89046-490 89046-506 89046-508 89046-510 89046-526 89046-528 89046-530 89046-542 89046-544 89046-556 89046-558 89046-560 89046-562 89046-564 89046-566 89046-568 89046-570 89046-572 89046-574 89046-576 89046-578 89046-580 89046-582 89046-584 89046-586 89046-588 89046-478 89046-480
AC Medium Rounds, Clear, Wide Mouth, Graduated, Kimble®, DWK Life Sciences
Bottles Glass Bottles
Versatile storage bottles provide an ideal containment for solid, liquid, or powdered specimens and samples.
The clear Type III soda-lime glass construction allows for constant sample viewing. Bottles feature graduated markings in both milliliters and ounces, and a wide mouth design for efficient sample addition and removal. Bottles are supplied with screw-on caps, unless otherwise noted.
Ordering information: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Corrugated cartons consist of shrink-wrapped glasswareMore Product Information