PST® 565™ Thread Sealant, Loctite®, Henkel
Supplier: Henkel
PST® 565™
88551 88552
19807-552CS 1425.37 CAD
19807-552 19807-550
PST® 565™ Thread Sealant, Loctite®, Henkel
Adhesives
PST® 565™ cures rapidly to provide low-pressure sealing for metal tapered pipe thread fittings.
  • Cures rapidly
  • Allows for ease of disassembly
  • Withstands continuous temperatures to 149°C (300°F)
Sealant's controlled strength allows for ease of disassembly. Sealant is solvent-resistant, protects mated threaded areas from rust and corrosion, and withstands continuous temperatures to 149°C (300°F). It takes four hours to seal to operating pressure.
Certifications: UL listed. AGA approved. NSF/ANSI 61, CFIA, and CSA certified.