AC Medium Rounds, Clear, Wide Mouth, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
5710133V-21 5710238V-21 5710448V-21 5710858V-21 5711670V-21 5710133C-21 5710238C-21 5710448C-21 5710858C-21 5711670C-21 5710238V-22 5710448V-22 5710133C-22 5710238C-22 5710448C-22 5710858C-22 5711670C-22 5710238V-25 5710133C-25 5710238C-25 5710133C-26 5710238C-26 5710448C-26 5710858C-26 5711670C-26 5710133B 5710238B 5710448B 5710858B 5711670B
89046-344CS 155.86 CAD
89046-344 89046-346 89046-348 89046-350 89046-352 89046-354 89046-356 89046-358 89046-360 89046-362 89046-366 89046-368 89046-374 89046-376 89046-378 89046-380 89046-382 89046-426 89046-428 89046-430 89046-442 89046-444 89046-446 89046-448 89046-450 89046-452 89046-454 89046-456 89046-458 89046-460
AC Medium Rounds, Clear, Wide Mouth, Kimble®, DWK Life Sciences
Bottles Glass Bottles
Versatile storage bottles provide an ideal containment for solid, liquid, or powdered specimens and samples.
The clear Type III soda-lime glass construction allows for constant sample viewing. Wide mouth design ensures efficient sample addition and removal. Bottles are supplied with screw-on caps, unless otherwise noted.
Ordering information: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 432 per wrap) packaged in corrugated cases. Bottle caps are included in separate bags within the case. Bottles sold without caps are also packaged in bulk packs.