Packers, Amber, Wide Mouth, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
5220128V-21 5220233V-21 5220438V-21 5220845V-21 5221653V-21 5223253V-21 5220128C-21 5220233C-21 5220438C-21 5220845C-21 5221653C-21 5223253C-21 5220128C-22 5220233C-22 5220438C-22 5220845C-22 5221653C-22 5223253C-22 5220845C-23 5221653C-23 5223253C-23 5220128V-25 5220233V-25 5220438V-25 5220128V-26 5220233V-26 5220438V-26 5220845V-26 5221653V-26 5223253V-26 5220128C-26 5220233C-26 5220438C-26 5220845C-26 5221653C-26 5223253C-26 5220128B 5220233B 5220438B 5220845B 5223253B
89045-596CS 87.2 CAD
89045-596 89045-598 89045-600 89045-602 89045-604 89045-606 89045-608 89045-610 89045-612 89045-614 89045-616 89045-618 89045-632 89045-634 89045-636 89045-638 89045-640 89045-642 89045-662 89045-664 89045-666 89045-692 89045-694 89045-696 89045-704 89045-706 89045-708 89045-710 89045-712 89045-714 89045-716 89045-718 89045-720 89045-722 89045-724 89045-726 89045-728 89045-730 89045-732 89045-734 89045-736 89045-738
Packers, Amber, Wide Mouth, Kimble®, DWK Life Sciences
Bottles Glass Bottles
Bottles are constructed of sturdy Type III soda-lime glass and feature a wide mouth design to aid in the addition and removal of sample contents.
The amber glass design provides protection for light-sensitive samples. Unless otherwise noted, bottles are supplied with black phenolic caps.
Packaging: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 432 per wrap) packaged in corrugated cases. Bottle caps are included in separate bags within the case. Bottles sold without caps are also packaged in bulk packs. Pallet packs consist of shrink-wrapped glassware bundles stacked onto shrink-wrapped pallets. This packaging is designed for large glassware quantities.