The Rapid Response™ Nitazene Test Strip (liquid/powder) detects the presence nitazene in liquid or powder samples at a cut-off concentration of 2000 ng/ml or higher.
- Sample: Liquid/powder
- Detects nitazene at concentrations of 2000 ng/ml and above
- Results as soon as 60 seconds
- Results interpretation guide printed on each pouch
- Every pouch features a QR code linking it to the BTNX Harm Reduction Resource Page - giving every user access to written and video instructions
- Easy to interpret results with built-in procedural controls
- Individually packaged strips for easy distribution
Warning: This test does not evaluate drug safety or purity.
The Rapid Response™ Nitazene Test Strip (Liquid/Powder) is an easy-to-use rapid lateral flow assy for the qualitative detection of Nitazene in liquid or powder dope samples.
It is specific for nitazene screening and can detect nitazene analogues including Isotonitazene, Protonitazene, and N-Pyrrolidino Etonitazene.
This kit includes individually packaged test strips with result interpretation guide printed on the back of each pouch, a result interpretation card to aid in reading the results, and detailed instructions.
To perform the test, remove the test strip from the pouch and hold the strip at the end where the product name is printed. Dip the test strip into liquid or powder sample (diluted in water) for 10-15 seconds. Remove the strip from the sample and place it on a non-absorbent flat surface. A negative result can be interpreted as soon as both the test (T) and control (C) lines appear. A positive result can be interpreted once the control line has appeared, there is no test line, and the background has cleared to white, typically by 60 seconds. Do not read results after 10 minutes. Please read the Product Insert supplied with the kit before use.
Ordering information: A timer is required to perform this test but is not provided in the kit. The kit should be stored at 2 to 30 °C (36 to 86 °F).
Packaging: Individually pouched test strips per box.
Caution: Not an IVD. For forensic use only. Do not use after expiration date indicated on the packaged. Do not reuse tests. Read the entire procedure carefully prior to performing any tests.
The test device is not intended to determine the purity, composition, or if the substance being examined is safe to use. A positive or negative test result is not an indication that the substance being examined is safe to use. Many factors come into play when examining the samples, including but not limited to mixture of multiple substances, solubility, and pH of the sample. BTNX Inc. does not encourage the use, supply, or production of illegal drugs or controlled substances in any way. The device is intended for harm reduction purposes. Follow the advice of your local harm reduction or public health agency. There is a possibility that technical or procedural errors as well as other substances and factors may interfere with the Rapid Response™ Nitazene Test Strip (Liquid / Powder) and cause false results. A positive result indicates the presence of nitazene only and does not indicate quantity. A negative result does not at any time rule out the presence of nitazene, as they may be present below the minimum detection level of the test.