BioXp® Select DNA Cloning Golden Gate Assembly BsmBI Kits
Supplier: TELESIS BIO
BX-SEL-CLNG10B-08 BX-SEL-CLNG10B-24 BX-SEL-CLNG4B-08 BX-SEL-CLNG4B-24
77506-790EA 255.33 CAD
77506-790 77506-792 77506-798 77506-800
BioXp® Select DNA Cloning Golden Gate Assembly BsmBI Kits
Nucleic Acid Reagents Cloning Kits and Ligation
The Select DNA cloning Golden Gate Assembly BsmBI kits are compatible with the Type IIS restriction enzymes BsmBI. Please choose the appropriate kit based on the number of fragments being assembled.
  • Accepted input: Telesis Bio DNA fragments, IDT gBlocks™, Twist Biosciences fragments
  • Supports user-provided linearized vector
  • Length: 0.3 to 1.8 kb
  • Supports up to >5 fragment assembly using BsmBI
  • Compatible with BioXp® 3250
The Select DNA cloning Golden Gate Assembly kits are compatible with the Type IIS restriction enzyme BsmBI. The user will need to select the number of fragments they want to assemble, either 1-4 fragment assembly or 5 fragment assembly.