BioXp® Select DNA Cloning Golden Gate Assembly, BsaI Kits
Supplier: TELESIS BIO
BX-SEL-CLNG10A-08 BX-SEL-CLNG10A-24 BX-SEL-CLNG4A-08 BX-SEL-CLNG4A-24
77506-786EA 255.33 CAD
77506-786 77506-788 77506-794 77506-796
BioXp® Select DNA Cloning Golden Gate Assembly, BsaI Kits
Nucleic Acid Reagents Cloning Kits and Ligation
The Select DNA cloning golden gate assembly, BsaI kits are compatible with the Type IIS restriction enzymes BsaI. Please choose the appropriate kit based on the number of fragments being assembled.
  • Accepted input: Telesis Bio DNA fragments, IDT gBlocks™, Twist Biosciences fragments
  • Supports user-provided linearized vector
  • Length: 0.3 to 1.8 kb
  • Supports up to >5 fragment assembly using BsaI
  • Compatible with BioXp® 3250
The Select DNA cloning golden gate assembly kits are compatible with the Type IIS restriction enzyme BsaI. The user will need to select the number of fragments they want to assemble, either 1 to 4 fragment assembly or 5 fragment assembly.