BioXp® Select DNA Cloning Gibson Assembly Kits
Supplier: TELESIS BIO
BX-SEL-CLNGA-08
BX-SEL-CLNGA-24
77506-808EA
255.33
CAD
77506-808
77506-810
BioXp® Select DNA Cloning Gibson Assembly Kits
Nucleic Acid Reagents
Cloning Kits and Ligation
The Select DNA cloning Gibson assembly kits can generate Gibson clones starting with fragments and vector supplied by the end user.
- Accepted input: Telesis Bio DNA fragments, IDT gBlocks™, Twist Biosciences fragments
- Supports user-provided linearized vector
- Length: 0.3 to 6.0 kb
- Compatible with BioXp® 3250
The Select DNA cloning Gibson assembly kits can generate Gibson clones starting with fragments and vector supplied by the end user.