BioXp® Select DNA Cloning and Amplification Kits
Supplier: TELESIS BIO
BX-SEL-CLNAMP-08 BX-SEL-CLNAMP-16 BX-SEL-CLNAMP-24
77506-682EA 957.48 CAD
77506-682 77506-684 77506-686
BioXp® Select DNA Cloning and Amplification Kits
Nucleic Acid Reagents Cloning Kits and Ligation
The Select DNA cloning and amplification kits generate a minimum of 10 µg of amplified DNA from fragments of 0.3 to 1.8 kb using the BioXp® 3250.
  • Accepted input: Telesis Bio DNA fragments, IDT gBlocks™, Twist Biosciences fragments
  • Supports user-provided linearized vector
  • Length: 0.3 to 1.8 kb
  • Yield: >10 µg/well
  • Compatible with BioXp® 3250
The BioXp® Select DNA cloning and amplification kit contains Gibson Assembly® and other reagents necessary to synthesize at least 10 µg of amplified DNA from fragments and linearized vectors supplied by the user.