Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (R-PE)

Supplier: Biotium

P012-RPE-125 P012-RPE-500
77449-098EA 102 CAD
77449-098 77449-100
Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (R-PE)
Antibodies
CD45 (clone 2D1) is a mouse monoclonal antibody that recognizes transmembrane glycoprotein CD45. This antibody have been validated and optimized in-house for optimal performance.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: 2D1
Conjugation: R-PE
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR