Anti-CD81 Mouse Monoclonal Antibody [clone: 5A6] (RPE (R-Phycoerythrin))

Supplier: Biotium

P005S-R616-125 P005S-R616-500
77449-074EA 317.26 CAD
77449-074 77449-076
Anti-CD81 Mouse Monoclonal Antibody [clone: 5A6] (RPE (R-Phycoerythrin))
Antibodies
CD81 (clone 5A6) is a validated mouse monoclonal antibody that recognizes the human tetraspanin protein CD81. The antibody belongs to the Biotium Choice list of select antibodies that have been validated and optimized in-house for optimal performance.

Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: 5A6
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Cynomolgus monkey, Human, Mouse (low reactivity), Non-human primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR