Durx® 670 Wipers, Berkshire
Supplier: Berkshire
DR670.082410 DR6700404.10 DR6700707.45 DR6700909.20 DR6701212.20 DR6701818.10
94000-404CS 730.02 CAD
94000-404 21913-410 21913-412 21913-431 21913-453 21913-475
Durx® 670 Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ideal for absorbing spills of solvents and deionized water in the cleanroom.
  • Excellent combination of synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Smooth, highly absorbent, and durable
  • Low extractable levels and particle counts
  • Chemically compatible with common cleaning and disinfecting solutions
  • Autoclavable
Also for general surface and equipment cleaning. Non-woven wipers are made from a polyester/cellulose blend of superior strength and absorbency, with low levels of ions and organic extractables.More Product Information