Standard Jars, Clear, Wide Mouth, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
5310448V-21 5310858V-21 5310448V-26 5310858V-26 5310448C-26 5310858C-26
89046-296CS 103.64 CAD
89046-296 89046-298 89046-328 89046-330 89046-332 89046-334
Standard Jars, Clear, Wide Mouth, Kimble Chase, DWK Life Sciences
Jars General Use Jars
Standard jars are manufactured from Type III soda-lime glass
The clear glass design features rounded shoulders and permits easy viewing of contents. Wide-mouth openings allow for easy addition and removal of contents. Jars are supplied with screw-on caps unless otherwise noted.
Ordering information: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 432 per wrap) packaged in corrugated cases. Bottle caps are included in separate bags within the case.
Order Now
SPECIFICATIONS
Environmentally Preferable