PIG® Aggressive Chemical Absorbent Packaging Bag
Supplier: New Pig
PKG3017 PKG3018 PKG3019 PKG3020 PKG3021
77463-562BX 119.48 CAD
77463-562 77463-564 77463-566 77463-568 77463-570
PIG® Aggressive Chemical Absorbent Packaging Bag
Bags Storage Bags
Secure your shipment from escaping aggressive chemicals by sealing your container in this packaging bag with absorbent sheet.
  • Sealable plastic bag with absorbent sheet traps and soaks up liquid escaping from container
  • Soft-but-heavyweight sheet adds light cushioning and scratch protection
  • Built-in mat flap lets you fold mat over the top for added barrier from seal point; tab also makes it easy to find the opening
  • Absorbs up to 1.41 ounces per bagged absorbent
  • Absorbent-bag combination works on most free aggressive liquids, including acids, bases and unknowns
  • Durable multi-layer construction holds together, even when saturated