DuraFrame® Rigid PCR Plates
Supplier: SCIENTIFIC SPECIALTIES, INC.
3730-00S 3730T00S 3735-00S 3735-01S 3735-02S 3735-04S 3735-05S 3735-06S 3735-09S 3735T00S 3742-00S 3742-01S 3742-02S 3742-04S 3742-05S 3742-06S 3742-09S 3742T00S 3750-00S 3750-01S 3750-02S 3750-04S 3750-05S 3750-06S 3750-09S
77506-814CS 1037.46 CAD
77506-814 77506-816 77506-818 77506-820 77506-822 77506-824 77506-826 77506-828 77506-830 77506-832 77506-838 77506-840 77506-842 77506-844 77506-846 77506-848 77506-850 77506-852 77506-858 77506-860 77506-862 77506-864 77506-866 77506-868 77506-870
DuraFrame® Rigid PCR Plates
Microplates PCR Plates
DuraFrame® plates feature a rugged polycarbonate frame, providing a solid and flat surface that guarantees seamless interaction with automated liquid handling platforms. With a firm grip assured, robots can confidently place DuraFrame® PCR plates with the utmost precision, eliminating any concerns of misalignment.
  • Polycarbonate frame is rigid and flat to facilitate a firm grip by robotic arms and ensure reliable placement
  • Clear polypropylene wells provide optimal thermal transfer and higher reaction efficiency
  • White well options enhance the fluorescence signal during qPCR (see 77506-856)
  • Wells are slightly raised to ensure evaporation-proof sealing with film, foil, or strip caps
  • ANSI/SBS footprint and well spacing
  • Vibrant colors offer a visual distinction between different samples or experiments
DuraFrame® plates feature polypropylene wells to facilitate optimal thermal transfer and higher reaction efficiency. This translates to more accurate and reliable results, giving researchers the confidence they need in their experiments. Furthermore, the white well options have been meticulously crafted to enhance the fluorescence signal during qPCR, enabling clearer and more precise measurements.
Sealing PCR plates is a crucial aspect of laboratory workflows, and DuraFrame® excels in this domain as well. The slightly raised wells of DuraFrame® PCR plates guarantee an evaporation-proof seal with film, foil, or strip caps. No more worries about sample integrity or loss due to evaporation - DuraFrame® ensures that valuable samples are protected throughout the entire process.
With an ANSI/SBS footprint and well spacing, DuraFrame® PCR plates seamlessly integrate into existing laboratory setups, offering compatibility and ease of use. Moreover, the vibrant color options provide a visual distinction between different samples or experiments, allowing for quick and efficient identification and organization.
Certifications: Certified free of RNase, DNase, DNA, and PCR inhibitors.
Packaging: Plates are packaged 10 per pack, 10 packs per case (100 plates in total).