TruCLEAN® Pro Cleaning and Disinfecting System, Perfex
Supplier: Perfex
22-15R 22-15W 22-15B 22-51 22-06B 22-06W 22-06R 22-3B 22-21 22-22B 22-22R 22-22W 22-22G 22-22Y 22-2R 22-2B 22-2W 22-3R 22-3W 22-20
22939-956EA 77.74 CAD
22939-956 22939-958 22939-960 22939-964 22939-966 22939-968 22939-970 22940-014 89096-002 89096-020 89096-022 89096-024 89096-026 89096-028 22940-006 22940-008 22940-010 22940-012 22940-015 89039-564
TruCLEAN® Pro Cleaning and Disinfecting System, Perfex
Mops
TruCLEAN® bucket systems are designed to capture and isolate contaminants, ensuring delivery of cleaning and sanitizing agents and reducing the risk of cross-contamination.
The double-bucket system includes stainless steel wringer, cart, 7.6cm (3") casters, handle holder, two 22L (5.8gal.) interlocking buckets, and mop frame with 86.4–157.5cm (34–62") polymer adjustable handle. The triple-bucket system includes stainless steel wringer, cart, 7.6cm (3") casters, handle holder, three 15L (4gal.) interlocking buckets, and mop frame with 86.4–157.5cm (34–62") polymer adjustable handle. The bucket underneath the wringer isolates soiled contaminates, while the front bucket holds cleaning and sanitizing agents. In the triple-bucket system, the middle bucket holds rinse to remove residual contaminates from the mop head.
Autoclavable, FDA-approved materials.
Optional stainless steel basket 22939-964 attaches to top of trolley and holds two 6L (1.6gal.) pails (22939-970, -968, -966) for carrying cleaning supplies. The polymer adjustable handle with red grip 22940-328 adjusts from 91.4–182.8cm (34–72") for use in hard-to-reach areas. The fixed mop frame 22940-326 eliminates swivel movement for hassle-free cleaning of ceilings and walls.
Ideal for quick, easy cleaning and sanitizing of large areas.
Ordering information: For other mop heads and replacement mop frames, see 22940-020 series.